Product Details

SNP ID
rs74593114
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:189661555 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGAGGGACGCGACCAGAGGACAGCT[C/T]TGTGCTGATCCCCACCGACAATTCG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ASNSD1 PubMed Links

Gene Details

Gene
ASNSD1
Gene Name
asparagine synthetase domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_019048.2 130 UTR 5 NP_061921.1
XM_017004381.1 130 Intron XP_016859870.1
XM_017004382.1 130 UTR 5 XP_016859871.1

View Full Product Details