Product Details

SNP ID
rs77652397
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:200519394 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAGATCTTACAGATACACAGCCATG[C/T]ATCACTTAAGGATGGGAATATGTTC
Phenotype
MIM: 612425
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KCTD18 PubMed Links

Gene Details

Gene
KCTD18
Gene Name
potassium channel tetramerization domain containing 18
There are no transcripts associated with this gene.

Gene
SGO2
Gene Name
shugoshin 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001160033.1 Intron NP_001153505.1
NM_001160046.1 Intron NP_001153518.1
NM_152524.5 Intron NP_689737.4
XM_005246344.3 Intron XP_005246401.2
XM_005246345.3 Intron XP_005246402.1
XM_011510734.2 Intron XP_011509036.1
XM_017003452.1 Intron XP_016858941.1
XM_017003453.1 Intron XP_016858942.1
XM_017003454.1 Intron XP_016858943.1

View Full Product Details