Product Details

SNP ID
rs77197458
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:219222049 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTCTCTTTTTTAGCTGTGCACTCA[C/G]CTGGTGCATATAGAGGGCATGCAGG
Phenotype
MIM: 605452 MIM: 612204
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ABCB6 PubMed Links

Gene Details

Gene
ABCB6
Gene Name
ATP binding cassette subfamily B member 6 (Langereis blood group)
There are no transcripts associated with this gene.

Gene
ANKZF1
Gene Name
ankyrin repeat and zinc finger domain containing 1
There are no transcripts associated with this gene.

Gene
ATG9A
Gene Name
autophagy related 9A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001077198.2 Intron NP_001070666.1
NM_024085.4 Intron NP_076990.4

View Full Product Details