Product Details

SNP ID
rs75993346
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:230722693 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCGTGTCTGATAGTGTGCTGTAAG[C/T]GTCCAATACATTTTAGCTTTTATGA
Phenotype
MIM: 612174
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CAB39 PubMed Links

Gene Details

Gene
CAB39
Gene Name
calcium binding protein 39
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001130849.1 Intron NP_001124321.1
NM_001130850.1 Intron NP_001124322.1
NM_016289.3 Intron NP_057373.1
XM_011511350.2 Intron XP_011509652.1

View Full Product Details