Product Details

SNP ID
rs506002
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:117837860 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATCCCTAAATCAGGGGAGGGGGCTG[G/T]TCTCGGGCAACAAGCAGCCTCCTAG
Phenotype
MIM: 601814 MIM: 606683
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
FXYD2 PubMed Links

Gene Details

Gene
FXYD2
Gene Name
FXYD domain containing ion transport regulator 2
There are no transcripts associated with this gene.

Gene
FXYD6
Gene Name
FXYD domain containing ion transport regulator 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001164831.2 1248 UTR 3 NP_001158303.1
NM_001164832.2 1248 UTR 3 NP_001158304.1
NM_001164836.2 1248 UTR 3 NP_001158308.1
NM_001164837.2 1248 UTR 3 NP_001158309.1
NM_022003.3 1248 UTR 3 NP_071286.1
Gene
FXYD6-FXYD2
Gene Name
FXYD6-FXYD2 readthrough
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001204268.1 1248 Intron NP_001191197.1
NM_001243598.2 1248 Intron NP_001230527.1

View Full Product Details