Product Details
- SNP ID
-
rs492799
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:65067834 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GGGTGGGTGGTGTGGGGGTGCCCAG[A/C]CAAAGGCCCCGGCTTCAGCAGGCCT
- Phenotype
-
MIM: 609374
MIM: 602640
- Polymorphism
- A/C, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
CDCA5
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs200668989,rs528283292] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CDCA5
- Gene Name
- cell division cycle associated 5
- Gene
- NAALADL1
- Gene Name
- N-acetylated alpha-linked acidic dipeptidase-like 1
There are no transcripts associated with this gene.
View Full Product Details