Product Details

SNP ID
rs35607512
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:163606652 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATAGATGGTCAGAATAGAGCTCAT[C/T]ATTAAGCTAGTTTATAGAAGAGCTC
Phenotype
MIM: 605295
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FIGN PubMed Links

Gene Details

Gene
FIGN
Gene Name
fidgetin, microtubule severing factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001321825.1 5495 UTR 3 NP_001308754.1
NM_018086.3 5495 UTR 3 NP_060556.2
XM_011511389.2 5495 UTR 3 XP_011509691.1
XM_017004419.1 5495 UTR 3 XP_016859908.1

View Full Product Details