Product Details

SNP ID
rs13013489
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:20670819 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCCGAGCCCGTTGGCACTGCGGCG[A/G]CTGGGCTTCGGCTGGCCGGGCGGAG
Phenotype
MIM: 604651
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GDF7 PubMed Links

Gene Details

Gene
GDF7
Gene Name
growth differentiation factor 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_182828.3 1323 Silent Mutation CGA,CGG R249R NP_878248.2

View Full Product Details