Product Details

SNP ID
rs11876682
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:70290712 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATCTGGAGTTTGGCCAGGGCTTTC[C/T]GCTTGTGCCTGGGTAAGGGTCAGCC
Phenotype
MIM: 605118
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SOCS6 PubMed Links

Gene Details

Gene
SOCS6
Gene Name
suppressor of cytokine signaling 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004232.3 Intron NP_004223.2
XM_005266783.3 Intron XP_005266840.1
XM_017026086.1 Intron XP_016881575.1
XM_017026087.1 Intron XP_016881576.1

View Full Product Details