Product Details

SNP ID
rs6975325
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:4778455 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TAGCTCGGGAGCTAGTGGGGCATGT[A/T]TCAGGCACAGGGTCTGTGCGAAGGG
Phenotype
MIM: 613653 MIM: 616302
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
AP5Z1 PubMed Links

Gene Details

Gene
AP5Z1
Gene Name
adaptor related protein complex 5 zeta 1 subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014855.2 Intron NP_055670.1
XM_017012864.1 Intron XP_016868353.1
Gene
FOXK1
Gene Name
forkhead box K1
There are no transcripts associated with this gene.

View Full Product Details