Product Details

SNP ID
rs13221118
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:157007928 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GACCATTTGGGAAATGCCATCCAAC[A/G]GGCAGAGACAACCAATTAAAGAAAC
Phenotype
MIM: 142994
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MNX1 PubMed Links

Gene Details

Gene
MNX1
Gene Name
motor neuron and pancreas homeobox 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001165255.1 Intron NP_001158727.1
NM_005515.3 Intron NP_005506.3
Gene
MNX1-AS1
Gene Name
MNX1 antisense RNA 1 (head to head)
There are no transcripts associated with this gene.

Gene
MNX1-AS2
Gene Name
MNX1 antisense RNA 2
There are no transcripts associated with this gene.

View Full Product Details