Product Details

SNP ID
rs2985
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:34877871 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCAATTATTCTGCTATTCCTGCCAT[C/T]ACCGCATCCTTCATGGTAGAGTATC
Phenotype
MIM: 600772
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC105375030 PubMed Links

Gene Details

Gene
LOC105375030
Gene Name
uncharacterized LOC105375030
There are no transcripts associated with this gene.

Gene
TAF11
Gene Name
TATA-box binding protein associated factor 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001270488.1 1859 UTR 3 NP_001257417.1
NM_005643.3 1859 UTR 3 NP_005634.1
XM_011514827.2 1859 UTR 3 XP_011513129.1
Gene
UHRF1BP1
Gene Name
UHRF1 binding protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017754.3 1859 Intron NP_060224.3

View Full Product Details