Product Details

SNP ID
rs15922
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:34883049 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TAATGATGAGTCTTCCCTTTCAACT[C/G]TTGTTAAATCTGAGACATCCTGACT
Phenotype
MIM: 608994 MIM: 600772
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ANKS1A PubMed Links

Gene Details

Gene
ANKS1A
Gene Name
ankyrin repeat and sterile alpha motif domain containing 1A
There are no transcripts associated with this gene.

Gene
LOC105375030
Gene Name
uncharacterized LOC105375030
There are no transcripts associated with this gene.

Gene
TAF11
Gene Name
TATA-box binding protein associated factor 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001270488.1 707 Missense Mutation ACA,AGA T68R NP_001257417.1
NM_005643.3 707 Missense Mutation ACA,AGA T68R NP_005634.1
XM_011514827.2 707 Missense Mutation ACA,AGA T24R XP_011513129.1
Gene
UHRF1BP1
Gene Name
UHRF1 binding protein 1
There are no transcripts associated with this gene.

View Full Product Details