Product Details

SNP ID
rs36113897
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.6:57052722 on Build GRCh38
Set Membership
Validated
Context Sequence [VIC/FAM]
TAAAATGCCAAAACGACAGGGTGAT[T/C]TTTTGCATTTTTTAAATGTGAAGAA
Phenotype
Polymorphism
T/C, Transition substitution
Allele Nomenclature
Literature Links
KIAA1586 PubMed Links

Gene Details

Gene
KIAA1586
Gene Name
KIAA1586
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286274.1 425 Missense Mutation CTT,TTT L48F NP_001273203.1
NM_001286275.1 425 Missense Mutation CTT,TTT L8F NP_001273204.1
NM_001286276.1 425 Missense Mutation CTT,TTT L8F NP_001273205.1
NM_020931.3 425 Missense Mutation CTT,TTT L75F NP_065982.1
XM_011514767.2 425 Missense Mutation CTT,TTT L75F XP_011513069.1

View Full Product Details