Product Details

SNP ID
rs11606478
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:86302628 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGAGGATGGAGCGTGGGAAGCTGG[C/G]TGCTCTCCTTTGCCCAGCTCTATTC
Phenotype
MIM: 614908
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
HIKESHI PubMed Links

Gene Details

Gene
HIKESHI
Gene Name
Hikeshi, heat shock protein nuclear import factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001322404.1 Intron NP_001309333.1
NM_001322407.1 Intron NP_001309336.1
NM_001322409.1 Intron NP_001309338.1
NM_016401.3 Intron NP_057485.2
XM_017017914.1 Intron XP_016873403.1
XM_017017915.1 Intron XP_016873404.1

View Full Product Details