Product Details

SNP ID
rs12828755
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:122986039 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACTCACGGTGCCCTACTTGGGGCC[C/T]GCGGCTGCCCCCGGCTCCAGGCGGC
Phenotype
MIM: 607668 MIM: 608920
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ARL6IP4 PubMed Links

Gene Details

Gene
ARL6IP4
Gene Name
ADP ribosylation factor like GTPase 6 interacting protein 4
There are no transcripts associated with this gene.

Gene
OGFOD2
Gene Name
2-oxoglutarate and iron dependent oxygenase domain containing 2
There are no transcripts associated with this gene.

Gene
PITPNM2
Gene Name
phosphatidylinositol transfer protein membrane associated 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001300801.1 4499 Silent Mutation NP_001287730.1
NM_020845.2 4499 Silent Mutation NP_065896.1
XM_011538590.2 4499 Silent Mutation XP_011536892.1

View Full Product Details