Product Details

SNP ID
rs11864713
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.16:15661888 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
AATTCATCAAGTTCTGGTAAGTTTG[C/G]CTTCTAAATATTTTTGGAGTCCATT
Phenotype
MIM: 609449
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
NDE1 PubMed Links

Gene Details

Gene
NDE1
Gene Name
nudE neurodevelopment protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001143979.1 Intron NP_001137451.1
NM_017668.2 Intron NP_060138.1
XM_005255396.4 Intron XP_005255453.1
XM_006720897.3 Intron XP_006720960.1
XM_006720900.3 Intron XP_006720963.1
XM_011522553.2 Intron XP_011520855.1
XM_017023349.1 Intron XP_016878838.1
XM_017023350.1 Intron XP_016878839.1
XM_017023351.1 Intron XP_016878840.1
XM_017023352.1 Intron XP_016878841.1
XM_017023353.1 Intron XP_016878842.1
XM_017023354.1 Intron XP_016878843.1
XM_017023355.1 Intron XP_016878844.1
XM_017023356.1 Intron XP_016878845.1
XM_017023357.1 Intron XP_016878846.1

View Full Product Details