Product Details

SNP ID
rs11556746
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:63326051 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAATCTCACACAGTTACAGCAAGCA[A/G]CTTTGAATTCCTGCTGCTTGGTCTG
Phenotype
MIM: 603263
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CA12 PubMed Links

Gene Details

Gene
CA12
Gene Name
carbonic anhydrase 12
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001218.4 1656 UTR 3 NP_001209.1
NM_001293642.1 1656 UTR 3 NP_001280571.1
NM_206925.2 1656 UTR 3 NP_996808.1

View Full Product Details