Product Details

SNP ID
rs11904760
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:110217118 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACCAGAGTCTTGAGTATGGGAAAAC[C/A]TTAGACACACAGAACTGGAAGAGAC
Phenotype
Polymorphism
C/A, Transversion substitution
Allele Nomenclature
Literature Links
LINC00116 PubMed Links

Gene Details

Gene
LINC00116
Gene Name
long intergenic non-protein coding RNA 116
There are no transcripts associated with this gene.

View Full Product Details