Product Details

SNP ID
rs7575835
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.2:110222769 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCACAGTCTCTCTCAGCAATGCAGA[G/A]TTGGTCTTGTAATACAGACTTCAGG
Phenotype
Polymorphism
G/A, Transition Substitution
Allele Nomenclature
Literature Links
LINC00116 PubMed Links

Gene Details

Gene
LINC00116
Gene Name
long intergenic non-protein coding RNA 116
There are no transcripts associated with this gene.

View Full Product Details