Product Details

SNP ID
rs4771120
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:27425574 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCTTGGGGTGGGAGGCAGGTGGGT[C/G]TGTTTGGGTGCGGAGATCACCATAT
Phenotype
MIM: 600860
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
GTF3A PubMed Links

Gene Details

Gene
GTF3A
Gene Name
general transcription factor IIIA
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002097.2 Intron NP_002088.2

View Full Product Details