Product Details

SNP ID
rs34769888
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:98199422 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCTCGTCCTCGCTGGCACAGCCCT[C/T]GGACACAGCTCTGCGCCGGGATGCC
Phenotype
MIM: 602159 MIM: 609871
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CORO2A PubMed Links

Gene Details

Gene
CORO2A
Gene Name
coronin 2A
There are no transcripts associated with this gene.

Gene
TBC1D2
Gene Name
TBC1 domain family member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001267571.1 2926 Missense Mutation AAG,GAG K916E NP_001254500.1
NM_001267572.1 2926 Missense Mutation AAG,GAG K456E NP_001254501.1
NM_018421.3 2926 Missense Mutation AAG,GAG K905E NP_060891.3
XM_011518843.1 2926 Missense Mutation AAG,GAG K456E XP_011517145.1
XM_011518844.2 2926 Intron XP_011517146.1
XM_017014899.1 2926 Missense Mutation AAG,GAG K471E XP_016870388.1

View Full Product Details