Product Details

SNP ID
rs4925151
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:18226818 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCCTCAGCCCACTCCTGGCTCACT[C/G]CTCTGGGGACGTTGGTTTTGTCCTC
Phenotype
MIM: 600966
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
LLGL1 PubMed Links

Gene Details

Gene
LLGL1
Gene Name
LLGL1, scribble cell polarity complex component
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004140.3 Intron NP_004131.3
XM_011523849.2 Intron XP_011522151.1
XM_011523850.2 Intron XP_011522152.1
XM_011523851.2 Intron XP_011522153.1
XM_011523852.2 Intron XP_011522154.1
XM_011523853.2 Intron XP_011522155.1
XM_011523854.2 Intron XP_011522156.1
XM_011523855.2 Intron XP_011522157.1
XM_011523856.1 Intron XP_011522158.1

View Full Product Details