Product Details

SNP ID
rs1219536
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:75720841 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTCTGGCTTTCCGACTCCTGTCACC[G/T]CGTGGTCATTGTGGACGCTGGGCCC
Phenotype
MIM: 610270
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
MOGAT2 PubMed Links

Gene Details

Gene
MOGAT2
Gene Name
monoacylglycerol O-acyltransferase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_025098.3 Intron NP_079374.2
XM_011545267.1 Intron XP_011543569.1

View Full Product Details