Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001009555.3 | Intron | NP_001009555.3 | |||
NM_001128923.1 | Intron | NP_001122395.1 | |||
NM_001128924.1 | Intron | NP_001122396.1 | |||
NM_001243349.1 | Intron | NP_001230278.1 | |||
XM_005262767.2 | Intron | XP_005262824.1 | |||
XM_011531646.1 | Intron | XP_011529948.1 | |||
XM_011531647.1 | Intron | XP_011529949.1 | |||
XM_011531648.1 | Intron | XP_011529950.1 | |||
XM_011531649.1 | Intron | XP_011529951.1 | |||
XM_017007778.1 | Intron | XP_016863267.1 | |||
XM_017007779.1 | Intron | XP_016863268.1 | |||
XM_017007780.1 | Intron | XP_016863269.1 | |||
XM_017007781.1 | Intron | XP_016863270.1 | |||
XM_017007782.1 | Intron | XP_016863271.1 | |||
XM_017007783.1 | Intron | XP_016863272.1 |