Product Details
- SNP ID
-
rs17628304
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
55
- Location
-
Chr.1:110454328 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- ATGGGGTTGCTGTGCCCCTGCCATC[A/C]ACAGGCATCCACCTTACTTAGTGCC
- Phenotype
-
MIM: 606233
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
PROK1
PubMed Links
Gene Details
- Gene
- PROK1
- Gene Name
- prokineticin 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_032414.2 |
|
Intron |
|
|
NP_115790.1 |
View Full Product Details