Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001204458.1 | Intron | NP_001191387.1 | |||
NM_001204459.1 | Intron | NP_001191388.1 | |||
NM_018647.3 | Intron | NP_061117.2 | |||
NM_148957.3 | Intron | NP_683760.1 | |||
XM_005266445.2 | Intron | XP_005266502.1 | |||
XM_005266446.1 | Intron | XP_005266503.1 | |||
XM_011535146.1 | Intron | XP_011533448.1 | |||
XM_017020651.1 | Intron | XP_016876140.1 | |||
XM_017020652.1 | Intron | XP_016876141.1 |