Product Details

SNP ID
rs3805395
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.4:168218519 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
TCACCTGCATGACTGTTACAGCCTT[C/T]GCACCTACATATTTCTGCCACATAT
Phenotype
MIM: 613974
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DDX60 PubMed Links

Gene Details

Gene
DDX60
Gene Name
DEXD/H-box helicase 60
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017631.5 Intron NP_060101.3
XM_011532103.2 Intron XP_011530405.1
XM_011532104.2 Intron XP_011530406.1
XM_017008383.1 Intron XP_016863872.1
XM_017008384.1 Intron XP_016863873.1

View Full Product Details