Product Details

SNP ID
hCV1861162
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:149039444 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATGCCCTGTGGCGTTTGGGGGGAG[C/G]TGGTGGGTGGAGGGGTGCTGTCCAC
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
LYPD6B PubMed Links

Gene Details

Gene
LYPD6B
Gene Name
LY6/PLAUR domain containing 6B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001317002.1 Intron NP_001303931.1
NM_001317003.1 Intron NP_001303932.1
NM_001317004.1 Intron NP_001303933.1
NM_001317005.1 Intron NP_001303934.1
NM_001317006.1 Intron NP_001303935.1
NM_177964.4 Intron NP_808879.2

View Full Product Details