Product Details

SNP ID
rs1535851
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.10:13224664 on Build GRCh38
Set Membership
Validated
Context Sequence [VIC/FAM]
AAGTGAAACTGGCATCCTCTGTGGC[A/G]ATGGCCAACAGGACCGCCTGGTGGC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RNU6-6P PubMed Links

Gene Details

Gene
RNU6-6P
Gene Name
RNA, U6 small nuclear 6, pseudogene
There are no transcripts associated with this gene.

Gene
UCMA
Gene Name
upper zone of growth plate and cartilage matrix associated
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001303118.1 Intron NP_001290047.1
NM_001303119.1 Intron NP_001290048.1
NM_145314.2 Intron NP_660357.2

View Full Product Details