Product Details

SNP ID
rs561756
Assay Type
Functionally tested
NCBI dbSNP Submissions
56
Location
Chr.1:52156489 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTGGTTTTGATTTACACCCAGGGGA[C/T]GGAGAGCTATTTTCAGAAGGTTTCA
Phenotype
MIM: 603755
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZFYVE9 PubMed Links

Gene Details

Gene
ZFYVE9
Gene Name
zinc finger FYVE-type containing 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004799.3 Intron NP_004790.2
NM_007324.3 Intron NP_015563.2
XM_011542437.2 Intron XP_011540739.1
XM_017002845.1 Intron XP_016858334.1

View Full Product Details