Product Details
- SNP ID
-
rs566864
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
49
- Location
-
Chr.1:119751125 on Build GRCh38
- Set Membership
-
JSNP
- Context Sequence [VIC/FAM]
- CTAGCACTTAAGGCCTTAATAAAAG[G/T]CTATTAAGACTTTTATTATAAGAGT
- Phenotype
-
MIM: 600234
MIM: 606879
- Polymorphism
- G/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
HMGCS2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs144498789] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- HMGCS2
- Gene Name
- 3-hydroxy-3-methylglutaryl-CoA synthase 2
- Gene
- PHGDH
- Gene Name
- phosphoglycerate dehydrogenase
There are no transcripts associated with this gene.
View Full Product Details