Product Details

SNP ID
rs1438537
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:69589932 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTTTAGCATAATTTTTCCAAAAAT[A/G]ATTTTGATACTGTGTGAGCTATACT
Phenotype
MIM: 604716
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
UGT2A1 PubMed Links

Gene Details

Gene
UGT2A1
Gene Name
UDP glucuronosyltransferase family 2 member A1 complex locus
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001252274.2 Intron NP_001239203.2
NM_001252275.2 Intron NP_001239204.2
NM_001301239.1 Intron NP_001288168.1
NM_006798.4 Intron NP_006789.3
Gene
UGT2A2
Gene Name
UDP glucuronosyltransferase family 2 member A2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001105677.2 Intron NP_001099147.2
NM_001301233.1 Intron NP_001288162.1

View Full Product Details