Product Details

SNP ID
rs2288740
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.4:69589861 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAATATATGCTTAACACAGTATGAT[C/G]AAATGTAGACATAAAAAAGTAGAGT
Phenotype
MIM: 604716
Polymorphism
C/G, Transversion Substitution
Allele Nomenclature
Literature Links
UGT2A1 PubMed Links
Additional Information
For this assay, SNP(s) [rs111934444] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
UGT2A1
Gene Name
UDP glucuronosyltransferase family 2 member A1 complex locus
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001252274.2 Intron NP_001239203.2
NM_001252275.2 Intron NP_001239204.2
NM_001301239.1 Intron NP_001288168.1
NM_006798.4 Intron NP_006789.3
Gene
UGT2A2
Gene Name
UDP glucuronosyltransferase family 2 member A2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001105677.2 Intron NP_001099147.2
NM_001301233.1 Intron NP_001288162.1

View Full Product Details