Product Details

SNP ID
rs1542021
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:200522972 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATTTCCTCAACTTTAAAAAGGAAAT[A/G]ATGGTACCTACCACATAGGATCATT
Phenotype
MIM: 612425
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SGO2 PubMed Links

Gene Details

Gene
SGO2
Gene Name
shugoshin 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001160033.1 Intron NP_001153505.1
NM_001160046.1 Intron NP_001153518.1
NM_152524.5 Intron NP_689737.4
XM_005246344.3 Intron XP_005246401.2
XM_005246345.3 Intron XP_005246402.1
XM_011510734.2 Intron XP_011509036.1
XM_017003452.1 Intron XP_016858941.1
XM_017003453.1 Intron XP_016858942.1
XM_017003454.1 Intron XP_016858943.1

View Full Product Details