Product Details
- SNP ID
-
rs3961919
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:112485744 on Build GRCh38
- Set Membership
-
HapMap
Validated
- Context Sequence [VIC/FAM]
- TGTGGGGAGAATGAAGGTTGCCAGC[A/G]GAATACTGAGAGAAAGAGGGTCCCT
- Phenotype
-
MIM: 608291
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
TTL
PubMed Links
Gene Details
- Gene
- TTL
- Gene Name
- tubulin tyrosine ligase
View Full Product Details