Product Details

SNP ID
rs7584726
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:19901602 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCGCTCAGTCCTAGTGCTGGGTTAT[A/G]ACAAGCGCCGACCGTGAGCTCCAGA
Phenotype
MIM: 613602
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TTC32 PubMed Links

Gene Details

Gene
TTC32
Gene Name
tetratricopeptide repeat domain 32
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001008237.2 Intron NP_001008238.1
Gene
WDR35
Gene Name
WD repeat domain 35
There are no transcripts associated with this gene.

View Full Product Details