Product Details
- SNP ID
-
rs10481332
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.8:23703319 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TACTGCTGGAACATAAATTGTGACA[A/C]CCTATGGTCCCTCGTGTGTGCGTCC
- Phenotype
-
MIM: 611770
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
NKX2-6
PubMed Links
Gene Details
- Gene
- NKX2-6
- Gene Name
- NK2 homeobox 6
View Full Product Details