Product Details

SNP ID
rs33929303
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:67911021 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCAGTTAAAAGGAGACCCTTGACAA[C/T]GACATTCTAAAAATCCCATGGACTA
Phenotype
MIM: 611704
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TMPRSS11A PubMed Links

Gene Details

Gene
TMPRSS11A
Gene Name
transmembrane protease, serine 11A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001114387.1 1699 UTR 3 NP_001107859.1
NM_182606.3 1699 UTR 3 NP_872412.3

View Full Product Details