Product Details
- SNP ID
-
rs28694577
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:153347356 on Build GRCh38
- Set Membership
-
Validated
- Context Sequence [VIC/FAM]
- GATTTCTCTGATTTGTCTCTTCAAG[C/T]CAGGCAATACATGATGTCACTGAAT
- Phenotype
-
MIM: 611422
MIM: 614141
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
MND1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs74875228] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- MND1
- Gene Name
- meiotic nuclear divisions 1
- Gene
- TRIM2
- Gene Name
- tripartite motif containing 2
There are no transcripts associated with this gene.
View Full Product Details