Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001195728.2 | 11490 | UTR 3 | NP_001182657.1 | ||
NM_001252652.1 | 11490 | UTR 3 | NP_001239581.1 | ||
NM_004171.3 | 11490 | UTR 3 | NP_004162.2 | ||
XM_005253067.1 | 11490 | UTR 3 | XP_005253124.1 | ||
XM_011520285.2 | 11490 | UTR 3 | XP_011518587.1 | ||
XM_017018136.1 | 11490 | UTR 3 | XP_016873625.1 | ||
XM_017018137.1 | 11490 | UTR 3 | XP_016873626.1 | ||
XM_017018138.1 | 11490 | UTR 3 | XP_016873627.1 | ||
XM_017018139.1 | 11490 | UTR 3 | XP_016873628.1 |