Product Details
- SNP ID
-
rs1418926
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.20:54210729 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CCAGGCCCCCAAAACTTTCTACTCC[G/T]AAGCTGTTTTATTTTCATCATCACG
- Phenotype
-
MIM: 604898
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
PFDN4
PubMed Links
Gene Details
- Gene
- PFDN4
- Gene Name
- prefoldin subunit 4
View Full Product Details