Product Details

SNP ID
rs1550824
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:16070511 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGCAAGTCAGTGATGGAGGGTGCAG[C/G]GGCACAGCTTCACCAGAGGAAACTA
Phenotype
MIM: 613338
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
MARCH11 PubMed Links

Gene Details

Gene
MARCH11
Gene Name
membrane associated ring-CH-type finger 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001102562.1 Intron NP_001096032.1

View Full Product Details