Product Details

SNP ID
rs179465
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:23763821 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCCGGGAGGAGGCTGGGCTGGCGC[C/G]CACTTCGTGCTTTGCAGGACTCAAG
Phenotype
MIM: 615903 MIM: 185261 MIM: 605017
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
C22orf15 PubMed Links

Gene Details

Gene
C22orf15
Gene Name
chromosome 22 open reading frame 15
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_182520.2 816 Intron NP_872326.2
XM_011529907.2 816 UTR 5 XP_011528209.1
XM_011529908.2 816 Intron XP_011528210.2
XM_011529912.2 816 UTR 5 XP_011528214.1
XM_017028602.1 816 UTR 5 XP_016884091.1
XM_017028603.1 816 UTR 5 XP_016884092.1
XM_017028604.1 816 UTR 5 XP_016884093.1
XM_017028605.1 816 UTR 5 XP_016884094.1
XM_017028606.1 816 UTR 5 XP_016884095.1
XM_017028607.1 816 UTR 5 XP_016884096.1
XM_017028608.1 816 UTR 5 XP_016884097.1
XM_017028609.1 816 Intron XP_016884098.1
XM_017028610.1 816 Intron XP_016884099.1
XM_017028611.1 816 UTR 5 XP_016884100.1
XM_017028612.1 816 Intron XP_016884101.1
XM_017028613.1 816 UTR 5 XP_016884102.1
XM_017028614.1 816 UTR 5 XP_016884103.1
XM_017028615.1 816 Intron XP_016884104.1
XM_017028616.1 816 Intron XP_016884105.1
XM_017028617.1 816 Intron XP_016884106.1
XM_017028618.1 816 Intron XP_016884107.1
Gene
CHCHD10
Gene Name
coiled-coil-helix-coiled-coil-helix domain containing 10
There are no transcripts associated with this gene.

Gene
MMP11
Gene Name
matrix metallopeptidase 11
There are no transcripts associated with this gene.

Gene
VPREB3
Gene Name
pre-B lymphocyte 3
There are no transcripts associated with this gene.

View Full Product Details