Product Details

SNP ID
rs203786
Assay Type
Functionally tested
NCBI dbSNP Submissions
42
Location
Chr.1:167928782 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATACCTTAATATTAGAATAAGCAGA[A/T]GTATAAACTAACAGATACCTGGCTA
Phenotype
MIM: 610494 MIM: 614737
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
DCAF6 PubMed Links

Gene Details

Gene
DCAF6
Gene Name
DDB1 and CUL4 associated factor 6
There are no transcripts associated with this gene.

Gene
MPC2
Gene Name
mitochondrial pyruvate carrier 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001143674.3 Intron NP_001137146.1
NM_015415.3 Intron NP_056230.1
XM_006711266.3 Intron XP_006711329.1

View Full Product Details