Product Details

SNP ID
rs1021487491
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:53534126 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTGCAGGGGTACCTTGGAAGTACC[C/T]GTGACAAACTGGAGGAACTTGGCAC
Phenotype
MIM: 300697
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
HUWE1 PubMed Links

Gene Details

Gene
HUWE1
Gene Name
HECT, UBA and WWE domain containing 1, E3 ubiquitin protein ligase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_031407.6 13306 Silent Mutation ACA,ACG T4301T NP_113584.3

View Full Product Details