Product Details
- SNP ID
-
hCV2226647
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.22:44569440 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ACCTTGAGGGAGGCGTGTGACATGG[A/G]AGTAAGTTTGCCAAAATTCATCAAA
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
LINC00207
PubMed Links
Gene Details
- Gene
- LINC00207
- Gene Name
- long intergenic non-protein coding RNA 207
There are no transcripts associated with this gene.
View Full Product Details