Product Details

SNP ID
rs236111
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:5952889 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TAAAAGAAGTTCTGAGGTTACCAGA[A/G]TGTGGGAATTCAAAGGTTGGAGAGG
Phenotype
MIM: 608187
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MCM8 PubMed Links

Gene Details

Gene
MCM8
Gene Name
minichromosome maintenance 8 homologous recombination repair factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001281520.1 Intron NP_001268449.1
NM_001281521.1 Intron NP_001268450.1
NM_001281522.1 Intron NP_001268451.1
NM_032485.5 Intron NP_115874.3
NM_182802.2 Intron NP_877954.1
XM_011529387.2 Intron XP_011527689.1
XM_017028105.1 Intron XP_016883594.1
XM_017028106.1 Intron XP_016883595.1
XM_017028107.1 Intron XP_016883596.1
Gene
TRMT6
Gene Name
tRNA methyltransferase 6
There are no transcripts associated with this gene.

View Full Product Details