Product Details

SNP ID
rs527459
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.9:35088145 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
ACATGACAGGAGTGCTGGGGTGAAC[A/G]GAGGGCACAAAACATACACATGAGA
Phenotype
MIM: 602956 MIM: 614730
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
FANCG PubMed Links

Gene Details

Gene
FANCG
Gene Name
Fanconi anemia complementation group G
There are no transcripts associated with this gene.

Gene
PIGO
Gene Name
phosphatidylinositol glycan anchor biosynthesis class O
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001201484.1 4238 Intron NP_001188413.1
NM_032634.3 4238 Intron NP_116023.2
NM_152850.3 4238 Intron NP_690577.2
XM_005251619.3 4238 UTR 3 XP_005251676.1
XM_017015222.1 4238 UTR 3 XP_016870711.1
XM_017015223.1 4238 Intron XP_016870712.1
XM_017015224.1 4238 Intron XP_016870713.1

View Full Product Details