Product Details

SNP ID
rs104894666
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:2250667 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCTCCGCAGAGCCTCTGCCCCTCC[C/T]GAGACACCCGCTACCTGGTGTTAGC
Phenotype
MIM: 600957 MIM: 608743 MIM: 600796
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
AMH PubMed Links

Gene Details

Gene
AMH
Gene Name
anti-Mullerian hormone
There are no transcripts associated with this gene.

Gene
JSRP1
Gene Name
junctional sarcoplasmic reticulum protein 1
There are no transcripts associated with this gene.

Gene
MIR4321
Gene Name
microRNA 4321
There are no transcripts associated with this gene.

Gene
SF3A2
Gene Name
splicing factor 3a subunit 2
There are no transcripts associated with this gene.

View Full Product Details